4-(3-Chlorophenoxy)benzoic acid ≥97%

Supplier: Matrix Scientific

113634-1G
102892-546EA 716.92 USD
102892-546
4-(3-Chlorophenoxy)benzoic acid ≥97%
4-(3-Chlorophenoxy)benzoic acid

Hazard: Irritant

Formula: C₁₃H₉ClO₃
MW: 248.67 g/mol
MDL Number: MFCD11122181
CAS Number: 1145-58-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR