5-Chloro-2-formylbenzoic acid ≥97%

Supplier: Matrix Scientific

113629-1G 113629-10G
102897-936EA 253.85 USD
102897-936 102897-938
5-Chloro-2-formylbenzoic acid ≥97%
5-Chloro-2-formylbenzoic acid

Hazard: Irritant

Formula: C₈H₅ClO₃
MDL Number: MFCD06739601
CAS Number: 4506-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR