1-(4-Methylphenyl)cyclopentanecarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(p-Tolyl)cyclopentanecarboxylic acid

113627-1G
102897-932EA 206.15 USD
102897-932
1-(4-Methylphenyl)cyclopentanecarboxylic acid ≥97%
1-(4-Methylphenyl)cyclopentanecarboxylic acid

Hazard: Irritant

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
MDL Number: MFCD00044672
CAS Number: 80789-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR