4-Bromo-3,5-dimethylbenzoic acid ≥97%

Supplier: Matrix Scientific

113623-1G
102897-922EA 1443.08 USD
102897-922
4-Bromo-3,5-dimethylbenzoic acid ≥97%
4-Bromo-3,5-dimethylbenzoic acid

Hazard: Irritant

Formula: C₉H₉BrO₂
MDL Number: MFCD16250111
CAS Number: 7697-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR