4-Ethylbenzenebutanoic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(4-Ethylphenyl)butanoic acid

113622-1G
102897-920EA 473.85 USD
102897-920
4-Ethylbenzenebutanoic acid ≥97%
4-Ethylbenzenebutanoic acid

Hazard: Irritant

Formula: C₁₂H₁₆O₂
MW: 192.26 g/mol
MDL Number: MFCD01462080
CAS Number: 5467-53-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR