3-Methoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: Matrix Scientific

113613-1G
102897-898EA 206.15 USD
102897-898
3-Methoxy-5-(trifluoromethyl)benzoic acid ≥97%
3-Methoxy-5-(trifluoromethyl)benzoic acid

Hazard: Irritant

Formula: C₉H₇F₃O₃
MW: 220.15 g/mol
MDL Number: MFCD09025409
CAS Number: 53985-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR