ɑ-Cyano-3-hydroxycinnamic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-3-(3-hydroxyphenyl)acrylic Acid

113610-1G
102892-540EA 135.38 USD
102892-540
ɑ-Cyano-3-hydroxycinnamic acid ≥97%
alpha-Cyano-3-hydroxycinnamic acid

Hazard: Irritant

Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00004203
CAS Number: 54673-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR