4-Butylbenzoic acid ≥97%

Supplier: Matrix Scientific

113609-1G
102892-538EA 60 USD
102892-538
4-Butylbenzoic acid ≥97%
4-Butylbenzoic acid

Hazard: Irritant

Formula: C₁₁H₁₄O₂
MW: 178.23 g/mol
MDL Number: MFCD00002571
CAS Number: 20651-71-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR