3-Furylmethylamine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Aminomethyl)furanFuran-3-ylmethanamine

113595-1G
102892-510EA 690.77 USD
102892-510
3-Furylmethylamine ≥97%
3-Furylmethylamine

Hazard: Irritant

Formula: C₅H₇NO
MW: 97.12 g/mol
MDL Number: MFCD02681983
CAS Number: 4543-47-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR