N-Methylfufurylamine ≥95%

Supplier: Matrix Scientific

Synonyms: N-Methylfurfurylamine

113593-10G 113593-1G
102892-506EA 946.15 USD
102892-506 102892-504
N-Methylfufurylamine ≥95%
N-Methylfufurylamine

Hazard: Irritant

Formula: C₆H₉NO
MW: 111.14 g/mol
MDL Number: MFCD00020961
CAS Number: 4753-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR