N-Methyl-(3-chloro-4-methoxy)benzylamine ≥97%

Supplier: Matrix Scientific

113590-1G
102892-498EA 958.46 USD
102892-498
N-Methyl-(3-chloro-4-methoxy)benzylamine ≥97%
N-Methyl-(3-chloro-4-methoxy)benzylamine

Hazard: Irritant

Formula: C₉H₁₂ClNO
MDL Number: MFCD09953609
CAS Number: 372117-85-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR