4-(1-Aminomethyl)benzonitrile ≥97%

Supplier: Matrix Scientific

113588-1G
102892-494EA 813.85 USD
102892-494
4-(1-Aminomethyl)benzonitrile ≥97%
4-(1-Aminomethyl)benzonitrile

Hazard: Irritant

Formula: C₉H₁₀N₂
MDL Number: MFCD09733813
CAS Number: 86225-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR