tert-Butyl-2-hydroxy-2-phenylethylcarbamate ≥97%

Supplier: Matrix Scientific

Synonyms: N-Boc-2-hydroxy-2-phenylethylamine

113582-1G
102892-482EA 641.54 USD
102892-482
tert-Butyl-2-hydroxy-2-phenylethylcarbamate ≥97%
tert-Butyl-2-hydroxy-2-phenylethylcarbamate

Hazard: Irritant

Formula: C₁₃H₁₉NO₃
MW: 237.3 g/mol
MDL Number: MFCD09750569
CAS Number: 67341-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR