5-Bromo-2-(2-ethylpiperidin-1-yl)benzaldehyde ≥97%

Supplier: Matrix Scientific

113559-1G
102892-434EA 738.46 USD
102892-434
5-Bromo-2-(2-ethylpiperidin-1-yl)benzaldehyde ≥97%
5-Bromo-2-(2-ethylpiperidin-1-yl)benzaldehyde

Hazard: Irritant

Formula: C₁₄H₁₈BrNO
CAS Number: 1229608-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR