tert-Butyl-2-pyridin-2-ylethylcarbamate ≥97%

Supplier: Matrix Scientific

113555-1G
102897-886EA 813.85 USD
102897-886
tert-Butyl-2-pyridin-2-ylethylcarbamate ≥97%
tert-Butyl-2-pyridin-2-ylethylcarbamate

Hazard: Irritant

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
MDL Number: MFCD03840873
CAS Number: 143185-43-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR