N-(2-Methoxyethyl)propylamine ≥97%
Supplier: Matrix Scientific
113552-1G
102897-880EA
156.92
USD
102897-880
N-(2-Methoxyethyl)propylamine ≥97%
N-(2-Methoxyethyl)propylamine
Purity: 97%
CAS: 43175-57-1
MDL Number: MFCD00144830
Molecular Formula: C6H15NO
Molecular Weight: 117,19
Hazard: Irritant
Formula:
C₆H₁₅NO MW: 117.19 g/mol |
MDL Number:
MFCD00144830 CAS Number: 43175-57-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...