N-(2-Hydroxyethyl)propionamide ≥97%
Supplier: Matrix Scientific
Synonyms:
N-Propionylethanolamine
113551-1G
102897-878EA
156.92
USD
102897-878
N-(2-Hydroxyethyl)propionamide ≥97%
N-(2-Hydroxyethyl)propionamide
Purity: 97%
CAS: 18266-55-2
MDL Number: MFCD00059600
Molecular Formula: C5H11NO2
Molecular Weight: 117,15
Hazard: Irritant
Formula:
C₅H₁₁NO₂ MW: 117.15 g/mol |
MDL Number:
MFCD00059600 CAS Number: 18266-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...