N-Cyclohexyl-N-cyclopropylamine ≥97%

Supplier: Matrix Scientific

113542-1G
102892-398EA 835.38 USD
102892-398
N-Cyclohexyl-N-cyclopropylamine ≥97%
N-Cyclohexyl-N-cyclopropylamine

Hazard: Irritant

Formula: C₉H₁₇N
MDL Number: MFCD09802453
CAS Number: 824-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR