6-Hydroxy-2,4,5-triaminopyrimidine ≥97%

Supplier: Matrix Scientific

Synonyms: 2,5,6-Triaminopyrimidin-4(1H)-one

113530-1G
102892-366EA 641.54 USD
102892-366
6-Hydroxy-2,4,5-triaminopyrimidine ≥97%
6-Hydroxy-2,4,5-triaminopyrimidine

TCA listed: T
Hazard: Irritant

Formula: C₄H₇N₅O
MW: 141.13 g/mol
Melting Pt: 242 °C
MDL Number: MFCD00217058
CAS Number: 1004-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR