tert-Butyl-N-(3-thienyl)carbamate ≥97%

Supplier: Matrix Scientific

Synonyms: tert-Butyl thiophen-3-ylcarbamate3-Aminothiophene, N-BOC protected

113527-1G
102892-360EA 544.62 USD
102892-360
tert-Butyl-N-(3-thienyl)carbamate ≥97%
tert-Butyl-N-(3-thienyl)carbamate

Hazard: Irritant

Formula: C₉H₁₃NO₂S
MW: 199.27 g/mol
MDL Number: MFCD01928808
CAS Number: 19228-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR