3-Amino-2-chlorophenol ≥97%

Supplier: Matrix Scientific

113520-1G
102892-322EA 583.08 USD
102892-322
3-Amino-2-chlorophenol ≥97%
3-Amino-2-chlorophenol

Hazard: Irritant/Store in Freezer

Formula: C₆H₆ClNO
MDL Number: MFCD11044694
CAS Number: 56962-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR