2,4,6-Trimethoxyaniline ≥97%

Supplier: Matrix Scientific

113518-1G
102892-318EA 447.69 USD
102892-318
2,4,6-Trimethoxyaniline ≥97%
2,4,6-Trimethoxyaniline

Hazard: Irritant

Formula: C₉H₁₃NO₃
MW: 183.21 g/mol
MDL Number: MFCD00017165
CAS Number: 14227-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR