N1,N1-Diethyl-4-[4-(diethylamino)benzyl]aniline ≥97%

Supplier: Matrix Scientific

Synonyms: 4,4'-Methylenebis(N,N-diethylaniline)

113516-1G
102892-312EA 641.54 USD
102892-312
N1,N1-Diethyl-4-[4-(diethylamino)benzyl]aniline ≥97%
N1,N1-Diethyl-4-[4-(diethylamino)benzyl]aniline

Hazard: Irritant

Formula: C₂₁H₃₀N₂
MW: 310.49 g/mol
MDL Number: MFCD00071778
CAS Number: 135-91-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR