2-Ethyl-5-nitroaniline ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Ethyl-5-nitrobenzenamine

113513-10G 113513-1G
102892-300EA 156.92 USD
102892-300 102892-302
2-Ethyl-5-nitroaniline ≥97%
2-Ethyl-5-nitroaniline

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.18 g/mol
MDL Number: MFCD00034056
CAS Number: 20191-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR