1-(5-Amino-2-hydroxyphenyl)ethanone ≥97%

Supplier: Matrix Scientific

113512-1G
102892-298EA 738.46 USD
102892-298
1-(5-Amino-2-hydroxyphenyl)ethanone ≥97%
1-(5-Amino-2-hydroxyphenyl)ethanone

Hazard: Irritant

Formula: C₈H₉NO₂
MW: 151.17 g/mol
MDL Number: MFCD00100397
CAS Number: 50-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR