3-Bromo-5-methoxyaniline ≥97%

Supplier: Matrix Scientific

113511-1G
102892-296EA 400 USD
102892-296
3-Bromo-5-methoxyaniline ≥97%
3-Bromo-5-methoxyaniline

Hazard: Irritant

Formula: C₇H₈BrNO
MDL Number: MFCD11878497
CAS Number: 16618-68-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR