2-Bromo-1-(4-fluoronaphthalen-1-yl)ethan-1-one ≥97%

Supplier: Matrix Scientific

113504-1G
102892-246EA 813.85 USD
102892-246
2-Bromo-1-(4-fluoronaphthalen-1-yl)ethan-1-one ≥97%
2-Bromo-1-(4-fluoronaphthalen-1-yl)ethan-1-one

Hazard: Irritant

Formula: C₁₂H₈BrFO
MDL Number: MFCD07787056
CAS Number: 388-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR