tert-Butyl-2,6-dimethyl-4-oxopiperidine-1-carboxylate

Supplier: Matrix Scientific

113485-1G
102892-240EA 1376.92 USD
102892-240
tert-Butyl-2,6-dimethyl-4-oxopiperidine-1-carboxylate
tert-Butyl-2,6-dimethyl-4-oxopiperidine-1-carboxylate
Formula: C₁₂H₂₁NO₃
MW: 227.31 g/mol
MDL Number: MFCD19230157
CAS Number: 604010-24-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR