6-Bromo-4-methylpyridin-2-amine
Supplier: Matrix Scientific
Synonyms:
2-Amino-6-bromo-4-methylpyridine
113480-1G
113480-5G
102892-234EA
553.85
USD
102892-234
102892-232
6-Bromo-4-methylpyridin-2-amine
6-Bromo-4-methylpyridin-2-amine
Hazard: Irritant
Formula:
C₆H₇BrN₂ MW: 187.05 g/mol |
MDL Number:
MFCD11036206 CAS Number: 73895-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...