MATR113474

113474-1G 113474-5G
102892-220EA 296.85 USD
102892-220 102892-222
MATR113474
CAS: 117976-89-3
MDL Number: MFCD00868879
Molecular Formula: C18H21N3O3S
Molecular Weight: 359,45
Hazard: Irritant
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR