Methyl cis-4-(boc-amino)cyclohexanecarboxylate

Supplier: Matrix Scientific

Synonyms: cis-Methyl 4-((tert-butoxycarbonyl)amino)cyclohexanecarboxylate

113469-1G 113469-5G
102897-856EA 170.77 USD
102897-856 102897-858
Methyl cis-4-(boc-amino)cyclohexanecarboxylate
Methyl cis-4-(boc-amino)cyclohexanecarboxylate

Hazard: Irritant

Formula: C₁₃H₂₃NO₄
MW: 257.33 g/mol
MDL Number: MFCD21641997
CAS Number: 364385-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR