4-Amino-1-methylcyclohexanol

Supplier: Matrix Scientific

113466-1G
102897-850EA 1740 USD
102897-850
4-Amino-1-methylcyclohexanol
4-Amino-1-methylcyclohexanol

Hazard: Irritant

Formula: C₇H₁₅NO
MDL Number: MFCD14706208
CAS Number: 233764-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR