(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate

Supplier: Matrix Scientific

Synonyms: (3S,4R)-tert-Butyl 3-fluoro-4-hydroxypiperidine-1-carboxylate

113458-1G
102892-210EA 1018.46 USD
102892-210
(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate
(3S,4R)-tert-Butyl-3-fluoro-4-hydroxypiperidine-1-carboxylate

Hazard: Irritant

Formula: C₁₀H₁₈FNO₃
MW: 219.26 g/mol
MDL Number: MFCD18632748
CAS Number: 1174020-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR