(S)-2-Aminomethyl-1-boc-azetidine

Supplier: Matrix Scientific

113437-1G
102897-844EA 1018.46 USD
102897-844
(S)-2-Aminomethyl-1-boc-azetidine
(S)-2-Aminomethyl-1-boc-azetidine

Hazard: Irritant

Formula: C₉H₁₈N₂O₂
MDL Number: MFCD17215571
CAS Number: 1007873-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR