6-Aza-spiro[3.4]octane oxalate

Supplier: Matrix Scientific

Synonyms: 6-Azaspiro[3.4]octane oxalate

113417-1G
102892-138EA 1890.77 USD
102892-138
6-Aza-spiro[3.4]octane oxalate
6-Aza-spiro[3.4]octane oxalate

Hazard: Irritant

Formula: C₉H₁₅NO₄
MW: 201.22 g/mol
MDL Number: MFCD23105788
CAS Number: 765-64-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR