4-Nitrooxindole

Supplier: Matrix Scientific

113408-1G
102892-096EA 1449.23 USD
102892-096
4-Nitrooxindole
4-Nitrooxindole

Hazard: Irritant

Formula: C₈H₆N₂O₃
MDL Number: MFCD09835635
CAS Number: 61394-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR