cis-3-Methoxycyclobutanamine hydrochloride

Supplier: Matrix Scientific

113397-1G
102892-068EA 1890.77 USD
102892-068
cis-3-Methoxycyclobutanamine hydrochloride
cis-3-Methoxycyclobutanamine hydrochloride

Hazard: Irritant

Formula: C₅H₁₂ClNO
MDL Number: MFCD22628709
CAS Number: 1408074-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR