3-Isoquinolinol
Supplier: Matrix Scientific
Synonyms:
3-Hydroxyisoquinoline
113373-5G
113373-1G
102891-996EA
401.54
USD
102891-996
102891-994
3-Isoquinolinol
3-Isoquinolinol
Hazard: Irritant
Formula:
C₉H₇NO MW: 145.16 g/mol |
MDL Number:
MFCD00075524 CAS Number: 7651-81-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...