(2R,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate hydrochloride
Supplier: Matrix Scientific
113362-1G
102891-978EA
1832.31
USD
102891-978
(2R,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate hydrochloride
(2R,5S)-tert-Butyl-2,5-dimethylpiperazine-1-carboxylate hydrochloride
Hazard: Irritant
Formula:
C₁₁H₂₃ClN₂O₂ |
MDL Number:
MFCD13191714 CAS Number: 792969-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...