(2S,5R)-1,2,5-Trimethylpiperazine oxalate

Supplier: Matrix Scientific

113360-1G
102891-974EA 2123.08 USD
102891-974
(2S,5R)-1,2,5-Trimethylpiperazine oxalate
(2S,5R)-1,2,5-Trimethylpiperazine oxalate

Hazard: Irritant

Formula: C₉H₁₈N₂O₄
MDL Number: MFCD26959166
CAS Number: 1152367-85-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR