trans-3-N-Boc-aminocyclopentanol

Supplier: Matrix Scientific

113333-1G
102891-948EA 920 USD
102891-948
trans-3-N-Boc-aminocyclopentanol
trans-3-N-Boc-aminocyclopentanol

Hazard: Irritant

Formula: C₁₀H₁₉NO₃
MDL Number: MFCD18791197
CAS Number: 207729-04-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR