2-Bromomethyl-5,8-dioxaspiro[3.4]octane

Supplier: Matrix Scientific

113314-1G
102897-738EA 1018.46 USD
102897-738
2-Bromomethyl-5,8-dioxaspiro[3.4]octane
2-Bromomethyl-5,8-dioxaspiro[3.4]octane

Hazard: Irritant

Formula: C₇H₁₁BrO₂
MDL Number: MFCD23106219
CAS Number: 1057641-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR