Ethyl-1H-pyrazolo[4,3-b]pyridine-6-carboxylate

Supplier: Matrix Scientific

113308-1G
102891-892EA 2181.54 USD
102891-892
Ethyl-1H-pyrazolo[4,3-b]pyridine-6-carboxylate
Ethyl-1H-pyrazolo[4,3-b]pyridine-6-carboxylate

Hazard: Irritant

Formula: C₉H₉N₃O₂
MDL Number: MFCD23106206
CAS Number: 1383735-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR