cis-4,4-Difluorooctahydrocyclopenta[c]pyrrole

Supplier: Matrix Scientific

113305-1G
102891-888EA 2560 USD
102891-888
cis-4,4-Difluorooctahydrocyclopenta[c]pyrrole
cis-4,4-Difluorooctahydrocyclopenta[c]pyrrole

Hazard: Irritant

Formula: C₇H₁₁F₂N
MDL Number: MFCD12198728
CAS Number: 1251008-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR