2-(Boc-aminomethyl)pyrimidine

Supplier: Matrix Scientific

Synonyms: tert-Butyl (pyrimidin-2-ylmethyl)carbamate

113294-1G
102891-870EA 1018.46 USD
102891-870
2-(Boc-aminomethyl)pyrimidine
2-(Boc-aminomethyl)pyrimidine

Hazard: Irritant

Formula: C₁₀H₁₅N₃O₂
MW: 209.25 g/mol
MDL Number: MFCD11617314
CAS Number: 1260843-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR