1-Boc-azepane-4-methylamine

Supplier: Matrix Scientific

113269-1G
102891-796EA 1687.69 USD
102891-796
1-Boc-azepane-4-methylamine
1-Boc-azepane-4-methylamine

Hazard: Irritant

Formula: C₁₂H₂₄N₂O₂
MDL Number: MFCD22035530
CAS Number: 1369353-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR