(R)-2-(Boc-aminomethyl)morpholine

Supplier: Matrix Scientific

Synonyms: tert-Butyl N-[(2R)-morpholin-2-ylmethyl]carbamate

113260-1G
102891-752EA 1890.77 USD
102891-752
(R)-2-(Boc-aminomethyl)morpholine
(R)-2-(Boc-aminomethyl)morpholine

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₃
MW: 216.28 g/mol
MDL Number: MFCD12545850
CAS Number: 186202-57-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR