cis-3-Methoxycyclobutanecarboxylic acid

Supplier: Matrix Scientific

113254-1G
102897-792EA 1629.23 USD
102897-792
cis-3-Methoxycyclobutanecarboxylic acid
cis-3-Methoxycyclobutanecarboxylic acid

Hazard: Irritant

Formula: C₆H₁₀O₃
MDL Number: MFCD22396943
CAS Number: 552849-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR