Methyl-3-oxo-1-methylcyclobutanecarboxylate

Supplier: Matrix Scientific

Synonyms: Methyl 1-methyl-3-oxocyclobutanecarboxylate

113243-1G
102891-734EA 607.69 USD
102891-734
Methyl-3-oxo-1-methylcyclobutanecarboxylate
Methyl-3-oxo-1-methylcyclobutanecarboxylate

Hazard: Irritant

Formula: C₇H₁₀O₃
MW: 142.16 g/mol
MDL Number: MFCD23105986
CAS Number: 1408075-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR