1-Boc-decahydro-1,5-naphthyridine

Supplier: Matrix Scientific

113221-1G
102891-688EA 2850.77 USD
102891-688
1-Boc-decahydro-1,5-naphthyridine
1-Boc-decahydro-1,5-naphthyridine

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₂
MDL Number: MFCD09863462
CAS Number: 1000931-58-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR